missing translation for 'onlineSavingsMsg'
Learn More

Kinematica™ Polytron™ PT 3100 D Homogenizer Drive Unit

Includes power cord, PC software, RS-232/USB connection cable

Brand:  Kinematica™ 11010122

Product Code. 13180673

  • 4350.00 € / Each
Estimated Shipment: 13-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Includes power cord, PC software, RS-232/USB connection cable
Specifications

Specifications

500 to 30,000 rpm
50/60 Hz
0.1 mL to 10 L
265 mm
130 mm
UK power cable, PC software, RS-232/USB connection cable
66 dBA at 25,000 rpm, 73 dBA at 30,000 rpm
0°C to 40°C
230 V
1200 W
240 x 130 x 265 mm
240 mm
LCD
PT 3100 D
UK Plug
Drive: 3.9 kg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Kinematica™ Polytron™ PT 3100 D Homogenizer Drive Unit >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.