missing translation for 'onlineSavingsMsg'
Learn More
Homoharringtonine, Tocris Bioscience™ Product Code.: 18712694

Homoharringtonine, Tocris Bioscience™

Product Code. 18712694
10 mg
Click to view available options
Quantity:
10 mg
This item is not returnable. View return policy

Product Code. 18712694

Brand: Tocris Bioscience 1416/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Inhibitor of protein synthesis; antileukemic agent

Inhibitor of protein synthesis. Blocks elongation phase of translation by binding to the 60-S ribosome subunit. Antileukemic.
TRUSTED_SUSTAINABILITY

Specifications

CAS 26833-87-4
Molecular Formula C29H39NO9
MDL Number MFCD05618221
Quantity 10 mg
Synonym dsstox_cid_25678, dsstox_rid_81052, dsstox_gsid_45678
InChI Key HYFHYPWGAURHIV-ZEDNPHJLSA-N
SMILES COC(=O)CC(O)(CCCC(C)(C)O)C(=O)O[C@H]1[C@@H]2C3=CC4=C(OCO4)C=C3CCN3CCC[C@]23C=C1OC
IUPAC Name (2R,3S,6R)-4-methoxy-16,18-dioxa-10-azapentacyclo[11.7.0.0²,⁶.0⁶,¹⁰.0¹⁵,¹⁹]icosa-1(20),4,13,15(19)-tetraen-3-yl 1-methyl 3-hydroxy-3-(4-hydroxy-4-methylpentyl)butanedioate
Molecular Weight (g/mol) 545.63
PubChem CID 45356872
Formula Weight 545.63
Percent Purity >99%
Chemical Name or Material Homoharringtonine
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Homoharringtonine, Tocris Bioscience™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.