missing translation for 'onlineSavingsMsg'
Learn More

Homopiperazine, 98%, Thermo Scientific Chemicals

Product Code. 11472398
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Homopiperazine is used to prepare 1,4-bis-(2-thiazolyl)-1,4-diazacycloheptane by reacting with 2-bromo-thiazole. It acts as an intermediate for repaglinide and fasudil hydrochloride. It is also used in the preparation of potent H3 receptor antagonists, which is used for the treatment of neurodegenerative conditions. Further, it is used in vesicant, cosmetic, emulsifier and energetic material. It acts as a corrosion inhibitor for iron. In pharmaceuticals, it is employed as antipyretic and reducing blood sugar, adiposity, calm, antipsychotic and anti-nervous.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Homopiperazine is used to prepare 1,4-bis-(2-thiazolyl)-1,4-diazacycloheptane by reacting with 2-bromo-thiazole. It acts as an intermediate for repaglinide and fasudil hydrochloride. It is also used in the preparation of potent H3 receptor antagonists, which is used for the treatment of neurodegenerative conditions. Further, it is used in vesicant, cosmetic, emulsifier and energetic material. It acts as a corrosion inhibitor for iron. In pharmaceuticals, it is employed as antipyretic and reducing blood sugar, adiposity, calm, antipsychotic and anti-nervous.

Solubility
Soluble in water.

Notes
Hygroscopic. Air sensitive. Incompatible with strong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 505-66-8
Molecular Formula C5H12N2
Molecular Weight (g/mol) 100.165
MDL Number MFCD00006933
InChI Key FQUYSHZXSKYCSY-UHFFFAOYSA-N
Synonym homopiperazine, 1,4-diazacycloheptane, 1h-1,4-diazepine, hexahydro, hexahydro-1,4-diazepine, 1,4 diazepane, perhydro-1,4-diazepine, trimethyleneethylenediamine, 1,4-diazepan, unii-95cl167w8t, hexahydro-1h-1,4-diazepine
PubChem CID 68163
IUPAC Name 1,4-diazepane
SMILES C1CNCCNC1

Specifications

Melting Point 38°C to 44°C
Density 0.95
Boiling Point 168°C to 170°C
Flash Point 64°C (147°F)
Odor Amine-like
Quantity 500 g
UN Number UN3259
Beilstein 102711
Sensitivity Air sensitive; Hygroscopic
Solubility Information Soluble in water.
Formula Weight 100.17
Percent Purity 98%
Chemical Name or Material Homopiperazine
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.