Learn More
Sperian™ Honeywell™ 4000 Premium Series Respirators
Reduces the risk of contamination and leakage, prevents irritation and allergies and increases safety. Sperian™ Honeywell™ 4000 Premium Series Respirators are easy to store and transport.
Brand:  Sperian™ 1005630
Product Code. 11831781
  • 165.00 € / Pack of 10
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Individually packaged, single use
  • With ultra-soft hypoallergenic Willtech™ flange seal
  • With absorption properties and a tight sealing film for exceptional comfort and fit for long periods
  • Preformed and colored adjustable nose bridge
  • Vast interior with a built-in chin piece
  • Ultra-lightweight with a fluid resistant outer shell
  • Sliding straps, highly flexible
TRUSTED_SUSTAINABILITY
Specifications
Specifications
EN 149:2001A1:2009, ISO 9001 / 2000, EC
FFP3
Medium
Textile, mining, construction, iron and steel industries, car body preparation industry, woodworking, agricultural and horticultural activities, food industry, cutting and molding of Metals, pharmaceutical manufacturing, nuclear industry, toxic waste treatment, battery, biological risks, etc
4311 ML Respirator
Dusts, Mists, Fumes
Single-use
Respiratory Protection
Yes
Yes
10/Pk.
Flat Fold