Learn More
Sperian™ Honeywell™ ARMAMAX AX™ Clear Lens Safety Goggles
Offering high levels of comfort and support. Sperian™ Honeywell™ ARMAMAX AX™ Clear Lens Safety Goggles fit comfortably over prescription eyewear.
12.00 € - 109.00 €
Specifications
Certifications/Compliance ISO 9001 / 2000, EN 166 1F
Lens Tint Clear
Product Type Eye and Face Protection Spectacles
For Use With (Application) Over-the-glass spectacle, General use and protection against impact (45m/s), Mechanic work: Grinding, turning, milling
Frame Color Transparent
View More Specs
Products 3
Product Code Brand Lens Coating Price Quantity & Availability  
Product Code Brand Lens Coating Price Quantity & Availability  
11847221
View Documents
Sperian™
1002221
Hardcoat This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12970405
View Documents
Sperian™
1002221
Hardcoat
12.00 €
Each
Estimated Shipment: 04-02-2025
to see stock.
Add to basket
11857221
View Documents
Sperian™
1002224
Uncoated
109.00 €
Pack of 10
In Stock
to see stock.
Edge
Add to basket
Description
Description
  • Practical and durable
  • Frosted side panels and browguard sections reduce glare/reflections
  • Curved temples and a raised nose bridge offer a precise fit
  • Available with or without hard lens coating
Specifications
Specifications
ISO 9001 / 2000, EN 166 1F
Eye and Face Protection Spectacles
Transparent
Polycarbonate
Clear
Over-the-glass spectacle, General use and protection against impact (45m/s), Mechanic work: Grinding, turning, milling
10/Pk.