Learn More
Sperian Protection™ Honeywell™ Optifit Series Silicone Full Face Mask
Full face mask designed for customized respiratory protection depending on the canister type assembled to it. Sperian Protection™ Honeywell™ Optifit Series Silicone Full Face Mask features a silicone skirt for comfort and resistance.
Brand:  Sperian Protection™ 1715021
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11887000
  • 255.00 € / Each
Estimated Shipment: 10-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Comfortable U-shaped silicone skirt
  • Optimal lateral and downward vision
  • Choice of filter
  • APR class 2 following EN 136:1998
TRUSTED_SUSTAINABILITY
Specifications
Specifications
EC Category PPE 3, Quality Assurance ISO 9001 / 2000, EC Certificate 0070-003-079-01-02-0009 - Ext. 01-10-03, EC Attestation Number 0070-003-079-01-02-0009 - Ext. 01-10-03, EN 136:1998
Full Face Mask
For Use with EN141 or EN143 Class 1 or 2 Certified Sperian Filters
Full Lens
Silicone
Large
RD40 connection
1 Ea.