Learn More
Sperian™ Honeywell™ FFP3 ML Respirator with Valve
Guarantees protection and wearer acceptance during periods of extended use. Sperian™ Honeywell™ FFP3 ML Respirators with Valve offers exceptional levels of comfort and no risk of allergies to latex.
Brand:  Sperian™ 1005602
Product Code. 11816970
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Rigid shell - maintains shape even in moist and heavy loading environments
  • Ultra-soft hypoallergenic Willtech™ flange seal with absorption properties and a tight sealing film for exceptional comfort and perfect fit
  • Preformed and colored nose bridge to save time adjusting the mask and instantly identify its protection level
  • Large internal volume to easily communicate and breathe
  • Shielded exhalation valve with an exclusive design for improved protection
  • Adjustable braids to easily and efficiently fit the mask
  • No latex, no silicone, no PVC
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Solid and/or Liquid
Single-use
Protects against dusts, mists and fumes containing calcium carbonate, clay, kaolin, cellulose, cotton, flour, ferrous Metals, cement, sodium silicate, sulphur, Glass fibers and plastics, hardwood, coal, quartz, non ferrous Metals, copper, aluminum, barium, titanium, vanadium, chrome, manganese, molybdenum, antimony, nickel, platinum, rhodium, strychnine, uranium, moderately radio-active particles, vegetal and mineral oils, Metal-working fluids (this list is non-exhaustive)
0.168 kg
EN 149:2001A1:2009
Respiratory Protection Molded Mask
5/Pk.