missing translation for 'onlineSavingsMsg'
Learn More

Honeywell™ Supervizor Face Shields

Product Code. p-8001516
Click to view available options
Model:
SB600 and SV9PC
SV9AC
SV9PC
Description:
200 mm Clear Acetate Visor
230 mm Clear Polycarbonate Visor
Browguard with Ratchet Headband
Browguard with Ratchet Headband and 230mm Clear Polycarbonate Visor
Lens Material:
Acetate
Polycarbonate
Unit Size:
Each
Pack of 10
This item is not returnable. View return policy

Product Code. 11836990

Brand: Honeywell™ 1002307

Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Robust, heavy-duty, helmet-mounted face shield range with hearing protection compatibility. Sperian™ Honeywell™ Supervizor Face Shields have a wide visor selection that can be tailored to the specific hazard/application.

  • Recommended for use in the chemical and forestry industries, among others
  • Protects against hazards such as abrasion, airborne particles, biohazards, chemicals, gases, vapors, and smoke
  • Maximum protection, allowing full unrestricted head movement
  • Creased visor eliminates gaps and visor profile follows contours of the face
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance EN 166 1F
Width (Metric) 200 mm
Description 200 mm Clear Acetate Visor
Product Type Supervisor Faceshield
Model SV9AC
No. per Pack 1 Ea.
Lens Material Acetate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.