missing translation for 'onlineSavingsMsg'
Learn More

Honeywell™ Valuair Half-Mask Air Purifying Respirator (APR)

Product Code. p-8010185
Click to view available options
Size:
Medium
Large
Unit Size:
Each
This item is not returnable. View return policy
For use only with certified T-Series cartridges (standards: EN 141, EN 143). The oxygen concentration must be superior to 17%.

Product Code. 12642653

Brand: Honeywell™ 1001574

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
For use only with certified T-Series cartridges (standards: EN 141, EN 143). The oxygen concentration must be superior to 17%.

Valuair Half-Mask APR features an elastomeric facepiece with soft, rounded edges and a low-profile nosepiece for low breathing resistance and eyewear compatibility. The twin filter uses a ‘click-fit’ system and the four-point harness offers secure positioning.

  • Material (Facepiece): Thermoplastic Elastomer
  • Harness Type: Four-Point Harness
  • Filter Type: Twin Filter System with Click-Fit Connection
  • Cartridge Type: P3 Dust Filter, A1 Gas Filter, A1P3 Combined Filter, A2 Gas Filter, A2P3 Combined Filter, ABE1 Gas Filter, ABE1P3 Combined Filter, ABEK1 Gas Filter, ABEK1P3 Combined Filter, K1 Gas Filter, K1P3 Combined Filter
  • Use with: Construction, Mechanical Engineering, Machining, Painting, Livestock and Horticulture
  • Standards/Compliance: EN 140, PPE Cat III, ISO 9001/2000
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO 9001/2000
Certifications/Compliance (Europe) PPE Cat. III
Certifications (EN) EN 140
Color Navy Blue
Design Type Low Profile
Disposable Reusable
Filter Type Twin Filter System with Click-Fit Connection
Material Thermoplastic Elastomer
Product Type Half-Mask Air Purifying Respirator (APR)
Size Large
Facepiece Material Thermoplastic Elastomer
For Use With (Application) Construction, Mechanical Engineering, Machining, Painting, Livestock and Horticulture
For Use With (Equipment) P3 Dust Filter, A1 Gas Filter, A1P3 Combined Filter, A2 Gas Filter, A2P3 Combined Filter, ABE1 Gas Filter, ABE1P3 Combined Filter, ABEK1 Gas Filter, ABEK1P3 Combined Filter, K1 Gas Filter, K1P3 Combined Filter
Harness Style Four-Point Harness
Quantity 1 Ea.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.