Learn More
Cole-Parmer™ ThermoSafe™ Compact Insulated Shipper
Choose from a variety of sizes to meet your refrigeration needs with these compact insulated shippers. Cole Parmer™ ThermoSafe™ Compact Insulated Shippers are ideal for shipping non-hazardous materials.
35.70 € - 47.30 €
Specifications
Type Compact Insulated Shipper
Quantity 1 Each
View More Specs
Products 2
Product Code Brand Dimensions (L x W x H) Price Quantity & Availability  
Product Code Brand Dimensions (L x W x H) Price Quantity & Availability  
11756028
View Documents
Cole-Parmer™
03730-20
6 x 6 x 6in.
47.30 €
Each
Estimated Shipment: 28-07-2025
to see stock.
Add to basket
11775509
View Documents
Cole-Parmer™
03730-30
8 x 8 x 11in. N/A N/A N/A
Description
Description
  • Compact shippers have a cylindrical interior to hold tubes, vials and other small items
  • Add as many ice packs as you need
  • For non-hazardous materials only
Specifications
Specifications
Compact Insulated Shipper
1 Each
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Cole-Parmer™ ThermoSafe™ Compact Insulated Shipper