Learn More
Medicom™ Hopen™ Single Use Hygiene Gown
Medicom™ Hopen™ Single Use Hygiene Gown without pockets, is microporous and waterproof
163.00 €
Specifications
Certifications/Compliance Manufactured under an ISO 9001 Certified Quality System
Certifications/Compliance (Europe) EN 1149-5
Color White
Cuff Style Elastic
Disposable Yes
View More Specs
Products 4
Product Code Brand Closure Type Size Price Quantity & Availability  
Product Code Brand Closure Type Size Price Quantity & Availability  
16524081
View Documents
Medicom™
40.550/02
Press studs Medium
163.00 €
Pack of 50
View alternative products
Save Up to 
»
Only null left
Estimated Shipment: 27-03-2026
Add to basket
16534081
View Documents
Medicom™
40.550/03
Press studs Large
163.00 €
Pack of 50
View alternative products
Save Up to 
»
Only null left
Estimated Shipment: 27-03-2026
Add to basket
16544081
View Documents
Medicom™
40.550/04
Press studs X-Large
163.00 €
Pack of 50
View alternative products
Save Up to 
»
Only null left
Estimated Shipment: 27-03-2026
Add to basket
16554081
View Documents
Medicom™
40.550/05
Press studs 2X-Large
163.00 €
Pack of 50
View alternative products
Save Up to 
»
Only null left
Estimated Shipment: 27-03-2026
Add to basket
Description
Description
  • Reduces contamination of the operator, but also sensitive products and processes in a protected atmosphere
  • Protects against radioactive particles and infectious agents
  • Microporous, Anti-Static Hygiene Gown is comfortable and light
  • Polypropylene gown lined with laminated polyethylene for a waterproof, hydrophobic seal
  • Available in mulitple sizes, and closure types
Specifications
Specifications
Manufactured under an ISO 9001 Certified Quality System
White
Yes
Category 1 Personal Protective Equipment
Polypropylene
Individual, Bag
Sewn
EN 1149-5
Elastic
Microporous, Waterproof
Unisex
None
Radioactive Particles, Infectious Agents
Non-sterile