missing translation for 'onlineSavingsMsg'
Learn More

Medicom™ Hopen™ Pocketed Hygiene Gown

Product Code. 16524061
Click to view available options
Color:
White
Size:
Medium
This item is not returnable. View return policy
This item is not returnable. View return policy

Medicom™ Hopen™ Pocketed Hygiene Gown with press-stud fasteners, available in two colors

  • Gown protects the operator, as well as sensitive products/processes that could be contaminated in a protected atmosphere
  • Protects user against radioactive particles, and infectious agents
  • Reduces the risk of soiling related to the user's activity
  • Pleasant and light, polypropylene fabric is breathable yet durable
  • Variety of colors allows for possibility of identification by color codes
  • Two pockets: one on the chest, and one on the lower thigh
  • Individually packaged for controlled hygiene, and distribution
  • Pockets located on the upper left, and lower right of the gown

Specifications

Size Medium
Construction Made in China
For Use With (Application) Category 1 Personal Protective Equipment
Gender Unisex
No. of Pockets 2
Packaging Type Individual, Bag
Product Type Hygiene Gown
Material Polypropylene
Color White
Closure Type Press Studs
Disposable Single-use
Sterility Non-sterile
Cuff Style Elastic
Seam Type Sewn
Resistance Infectious Agents
Certifications/Compliance Manufactured under an ISO 9001 Certified Quality System
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'