missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ratiolab™ Horizontal Cryo Racks for Freezer Cabinets
For the storing of Cryo Boxes in freezer cabinets and chests.
Brand: Ratiolab™ 5400009
This item is not returnable.
View return policy
Description
- Manufactured from stainless steel
- Surface is perfect for easy cleaning
- Robust construction for long life
- Handles for convenient transport
- Racks can be supplied with shelf sizes for any standard Cryo Box
- For all Cryo Boxes format 130 x 130 mm, 133 x 133 mm and 136 x 136 mm
- Supplied individually
Specifications
424 x 140 x 219 mm | |
For box 50 mm | |
3 x 4 | |
Horizontal rack |
Cryo box storage | |
1/pk | |
Stainless Steel |