Learn More
Fisherbrand™ Horizontal Large Gel Systems: Combs
For large gel systems
Brand:  Fisherbrand™ A2-8D
 View more versions of this product
Product Code. 12453657
  • 58.10 € / Each
Estimated Shipment: 28-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
For large gel systems
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Owl A2 and A5 Large Gel Electrophoresis System
25 x 20 cm
0.15 cm
9.8 in.
7.8 in.
20 cm
Standard Comb
8
165 μL
25 cm
0.88 in.
2.25 cm