missing translation for 'onlineSavingsMsg'
Learn More

Scientific Industries Horizontal Orbital Shaker Attachment for Roto-Shake Genie

An accessory that adds horizontal orbital shaking capability to the Roto-Shake Genie.

Brand:  Scientific Industries SI-1175

Product Code. 16647762

  • 366.00 € / Each
Estimated Shipment: 28-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Adds horizontal orbital shaking capability to the Roto-Shake Genie with variable speed of 15-100 rpm
  • Ideal for Western Blots and for maintaining cells or solutions in suspension
  • No tools required
  • Simply remove the Magnetic Platform from the Roto-Shaker Genie and attach the Horizontal Orbital Shaker Attachment
  • Fits all new and existing Roto-Shake models
Specifications

Specifications

2 kg
13 in.
13 x 13 in.
5 in.
13 in.
330.20 mm
1.6 kg
Horizontal Orbital Shaker Attachment for Roto-Shake Genie
Scientific Industries Roto-Shake Genie
1
Various containers, flasks and microplates
330.20 mm
13 x 13 x 5 in.
127 mm
330.20 mm
3.5 lb.
Black
Orbital Shaking of Various Containers Including Flasks and Microplates.
Horizontal Orbital Shaker Attachment with Texture Mat
1 Ea.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.