missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Horse anti-Mouse M.O.M.(R) (Mouse-On-Mouse) IgG Secondary Antibody, Biotin, Novus Biologicals™
Horse Polyclonal Antibody
Brand: Novus Biologicals MKB-2225-NB
This item is not returnable.
View return policy
Description
M.O.M.(R) (Mouse-On-Mouse) IgG Polyclonal antibody specifically detects M.O.M.(R) (Mouse-On-Mouse) IgG in Mouse samples. It is validated for Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), ImmunoblotSpecifications
M.O.M.(R) (Mouse-On-Mouse) IgG | |
Polyclonal | |
Equine | |
Affinity purified | |
RUO | |
Mouse | |
Purified |
Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot | |
Biotin | |
Mouse IgG | |
0.1 mL | |
Secondary | |
Store at 4°C. Do not freeze. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction