Learn More
Fisherbrand™ Hose Adapter Kit for Rotary Vane Vacuum Pumps
Use this hose adaptor kit with all Fisherbrand rotary van vacuum pumps. Fisherbrand™ Hose Adapter Kits for Rotary Vane Vacuum Pumps include stainless steel tubing adaptors and clamps for vacuum tight connections.
Brand:  Fisherbrand™ 330060
Product Code. 15576335
  • 335.00 € / Each
Estimated Shipment: 22-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Vacuum hose adapter kit
  • For Fisherbrand rotary vane vacuum pumps
  • Stainless steel tubing adaptors and clamps for vacuum tight connections
  • 1.5m of thick walled rubber hose in Sizes DN10 and DN20
Specifications
Specifications
DN10 & DN20 hose, Stainless Steel adapters and clamps
All Fisherbrand vacuum pumps
Hose Adapter Kit for Rotary Vane Vacuum Pumps