missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ Metal Stop-it Hose Clamp

Buerkle™ Metal Stop-it Hose Clamp can be used wherever the flow in hoses with liquids or gases needs to be regulated or stopped.

Brand:  Buerkle™ 8619-0520

6.43 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 11931601

  • 7.35 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Galvanised steel and aluminum
  • For flexible hoses with outer Ø up to 10mm, 15mm, 20mm and 30mm
  • Ensure that the stop-it metal is extremly heat resistant and sturdy
  • With the hose clip stop-it's knurled screw, a stepless, delicate flow regulation or complete stop of the flow are possible, even for existing hose connections
  • The hose is compressed over a wide area against the sturdy steel hook in order to prevent it from being damaged
Specifications

Specifications

Stop-It Hose Clamp
Galvanized Steel, Aluminum
20 mm O.D. Flexible Hoses
Extremely heat resistant and sturdy
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.