missing translation for 'onlineSavingsMsg'
Learn More

Kimberly-Clark™ Hostess™ Jumbo Roll Toilet Tissue
GREENER_CHOICE

Breaks down in cold water so it won’t block drains. Hostess™ Jumbo Roll Toilet Tissue helps to reduce maintenance and control costs and waste.

Brand:  Kimberly-Clark™ 8614

55.67 EUR valid until 2025-02-13
Use promo code "24130" to get your promotional price.


 View more versions of this product

Product Code. 12794246

  • 109.00 € / 12 rolls
Estimated Shipment: 18-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • White
  • Perforated, 2-ply tissue
  • Supplied in packs of 12 Jumbo Rolls x 500 Sheets
  • ISO 14001 and ISO 9001 compliant
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Toilet Tissue
9 cm
40 cm
ISO 14001 and ISO 9001, Ecolabel and FSC certified
40 x 9 cm
Ideal for busy washrooms in public venues and work environments, especially where economy is important
2-ply
8.03 in.
16.5 in.
Paper
White
76 mm core
12 x 200 m Roll
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.