missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Hot Gas Bypass Temperature Controllers

Combine these hot gas bypass temperature controllers with Thermo Scientific™ CryoPlus™ LN2 cryopreservation storage systems for increased utility.

3420.00 € - 3457.20 €

Specifications

For Use With (Equipment) CryoPlus 1,2,3,4
Voltage 120/220 V
View More Specs

Products 2
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
11674210
View Documents
Thermo Scientific™
195700
Field installed (requires qualified professional); 120V
3420.00 €
Each
Estimated Shipment: 20-03-2025
to see stock.
Add to basket
 
11767497
View Documents
Thermo Scientific™
195701
Field installed (requires qualified professional); 220V
3457.20 €
Each
In Stock
to see stock.
Add to basket
 
Description

Description

Combine Thermo Scientific™ Hot Gas Bypass Temperature Controllers with Thermo Scientific™ CryoPlus™ LN2 cryopreservation storage systems for increased utility.

Features:
  • 120V or 220V
  • Factory or field installed

Ordering Information:

Field installed option must be installed by a qualified professional.

  • Warranty varies by country, please contact us for details
  • Specifications

    Specifications

    CryoPlus 1,2,3,4
    120/220 V
    Videos
    SDS
    Documents

    Documents

    Product Certifications
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.