missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ HotStart™ Storage Reaction Tubes

Product Code. 15292559 Shop All Thermo Scientific Products
Click to view available options
Product Line:
Hot Start Micro 50 Bulk
HotStart Micro 50 Strips
Volume (Metric):
0.2 mL
Unit Size:
Pack of 120
Pack of 480
This item is not returnable. View return policy

Product Code. 15292559

Brand: Thermo Scientific™ 6210

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Eliminate tedious oil or wax overlays with the thin-walled Thermo Scientific™ HotStart™ Storage Reaction Tubes.

Features:
  • Bead is stable at room temperature
  • Assures synchronous reaction startup
  • PCR product can remain in tube for later analysis
  • Hinged caps provide snug seal
  • Presterilized; RNase- and DNase-free
Recommended for: Complex DNA or cDNA, very low copy number targets, more than 30 thermal cycles, multiple PCR or RT-PCR
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 0.2 mL
For Use With (Equipment) Thermal Cyclers
Product Line HotStart Micro 50 Strips
Sterility Sterile
Closure Type Flat Cap
Reaction Volume 25 to 50 μL
Product Type Strips
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.