missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Honeywell™ Howard Leight™ Leightning Ultraslim Neckband Earmuffs
Click to view available options
22 DB NRR; 1 pair
Description
- Robust steel wire construction withstands demanding use, especially in tough environments
- Features patented Air Flow Control technology, delivering optimal attenuation across all frequencies
- Snap-in ear cushions make replacement quick and easy
- Multiple attenuation levels allow for targeted attenuation for a variety of applications
- Telescopic height adjustment remains fixed during use
- Delivers high degree of comfort; ideal for situations where compact earmuffs and reliable Noise-Blocking protection are required
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Specifications
Color | Black |
Includes | Padded foam headband, snap-in ear cushions, telescopic height adjustment |
Certifications/Compliance | ANSI S3.19-1974 |
For Use With (Application) | Agriculture, construction, fire protection, food services, forestry, government, law enforcement, manufacturing, medical, military, municipal services, oil and gas, pharmaceutical, Steel and Metals transportation, welding, wind energy |
Noise Reduction Rating (NRR) | 22 dB |
Product Type | Ear Muffs |
Dielectric | No |
Style | Behind-the-neck |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction