missing translation for 'onlineSavingsMsg'
Learn More

Schmidlin™ HP Plus Rack Nitrogen Generator

This nitrogen generator produces a continuous supply of high purity nitrogen.

Brand:  Schmidlin™ DB-N2R-500-O-EU

Encompass_Preferred

Product Code. 17778889

  • 9873.56 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Increase efficiency and improve safety and performance
  • On-demand supply 24/7
  • Purity: <0.1 ppm THC
  • Pressure: up to 5 barg (75 psig)
  • Proprietary carbon molecular sieve technology
  • Ideal for analyzer applications
  • Stable baseline with increased sensitivity and reputability
  • Exceeds requirements for the most demanding applications
  • Superior nitrogen purification with long life catalyst
Specifications

Specifications

500 cm
13.3 cm
5 bar
48.5 cm
110/120 V at 60 Hz, 220/240 V at 50 Hz
270 W
500 mL/min.
35°C
19 in.
Outlet: 1/8 in.
>99.999%
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers