Learn More
Epredia™ HP35 Disposable Microtome Blades
Epredia™ HP35 microtome blades are coated for reliable and thin sectioning.
Brand:  Epredia™ 3150734
 View more versions of this product
Product Code. 11912355
  • 295.00 € / Pack of 50
Explore more special offers
Description
Description
  • High-profile
  • Increased durability
TRUSTED_SUSTAINABILITY
Specifications
Specifications
High Profile Blade
Designed for high-quality thin sectioning
Coated
0.01 in.
High
Single-use
35°
0.31 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Epredia™ HP35 Disposable Microtome Blades > Microtome blades; Coated