missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HPGDS Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22522-1-AP-150UL
This item is not returnable.
View return policy
Description
PGDS is a sigma class glutathione-S-transferase family member. The enzyme catalyzes the conversion of PGH2 to PGD2 and plays a role in the production of prostanoids in the immune system and mast cells. The presence of this enzyme can be used to identify the differentiation stage of human megakaryocytes. Prostaglandin-D synthase is a sigma class glutathione-S-transferase family member. The enzyme catalyzes the conversion of PGH2 to PGD2 and plays a role in the production of prostanoids in the immune system and mast cells. The presence of this enzyme can be used to identify the differentiation stage of human megakaryocytes.Specifications
HPGDS | |
Polyclonal | |
Unconjugated | |
HPGDS | |
Glutathione S transferase, GST class sigma, GSTS, H PGDS, HPGDS, PGDS, Prostaglandin H2 D isomerase, PTGDS2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
27306, 54486 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O60760, Q9JHF7 | |
HPGDS | |
HPGDS Fusion Protein Ag18290 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction