Learn More
Thermo Scientific™ dNTP Mix (20 mM)
Thermo Scientific dNTP Mix (20 mM) contains purified deoxyribonucleotide triphosphates (dNTPs) that meet high quality standards that result in excellent performance for a variety of molecular biology applications.
Brand: Thermo Scientific™ AB0196
52.16 EUR valid until 2025-03-21
Use promo code "24111" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price
Description
Thermo Scientific dNTP Mix (20 mM) contains purified deoxyribonucleotide triphosphates (dNTPs) that meet high quality standards that result in excellent performance for a variety of molecular biology applications. This pre-mixed solution contains 5 mM each of dATP, dCTP, dGTP, and dTTP at pH 7.5 for a total dNTP concentration of 20 mM. Such a mix makes it possible to reduce the total number of pipetting steps and the risk of reaction set-up errors.
Features
• Purity confirmed by HPLC
• Stable for 2 years at –20°C
• Free of human and E. coli DNA
Applications
Thermo Scientific dNTP mix may be used in a wide variety of molecular applications including:
• cDNA synthesis
• PCR, RT-PCR, qPCR
• Primer extension

Specifications
dNTP Mix (20 mM) contains premixed dATP, dCTP, dGTP and dTTP at a final concentration of 20 mM (1 mL) Store at –20°C. |
|
Unlabeled | |
Approved for shipment on Wet or Dry Ice | |
20 mM |
Liquid | |
1 mL | |
Standard PCR, Real Time PCR (qPCR) |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.