Learn More
SCAT™ HPLC Supply and Waste Set, V2.0, Suitable for All HPLC Systems
The all-round carefree package suitable for all HPLC and UHPLC systems.
836.00 € - 940.00 €
Specifications
For Use With (Equipment) Any HPLC System
View More Specs
Products 4
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
16458190
View Documents
SCAT™
307337
HPLC Supply and Waste Set, V2.0, suitable for all HPLC systems
836.00 €
Each
Estimated Shipment: 21-01-2025
to see stock.
Add to basket
17865704
View Documents
SCAT™
307447
845.00 €
1 set
Estimated Shipment: 23-01-2025
to see stock.
Add to basket
18626478
View Documents
SCAT™
307338
HPLC Supply and Disposal Set (12 Month), Non-Electrostatic Conductive
836.00 €
1 set
18636478
View Documents
SCAT™
307448
HPLC Supply and Disposal Set (12 Month), Electrostatic Conductive
940.00 €
1 sheet
Description
Description
  • Protects your system from contamination
  • Keeps solvent mixtures stable
  • High performance materials with excellent chemical resistance
The safety set connects tubing safely and blocks hazardous vapours. Air valves and exhaust filters provide safe pressure equalisation without under- or overpressure in solvent bottles and waste containers
Specifications
Specifications
Any HPLC System