Learn More
SCAT™ HPLC Waste Sets, V2.0
HPLC Waste Sets, V2.0 are economical combinations for clean and safe disposal of liquid laboratory waste by HPLC systems or other analytical instruments. All sets consist of a canister and an original SCAT™ SafetyCap with exhaust filter as well as fittings, blind plugs and a tube connector.
Brand:  SCAT™ 307373
331.76 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 17865714
  • 450.00 € / 1 set
Estimated Shipment: 18-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
SCAT™ offers a complete system for safe supply and disposal of solvents in analytics. All products are compatible across generations and perfectly matched to each other.
  • Customized system with high-quality set components
  • Safety canisters made of HDPE or polypropylene material
  • Attractive price compared to single purchase
  • Ready to use, no further components necessary
Specifications
Specifications
HPLC Systems
Safety Waste Cap, 2 x PFA fitting 2.3 mm OD, 2 x PFA fitting 3.2 mm OD, 1 x PP tube connector 5 to 11.5 mm ID, 1 x GL14 connection for exhaust filter, 1 x space saving canister, S50, 5 L, PE-HD elektrostatic conductive, with floater, 1 x grounding cable, 2 x clamp, length 1.5 m, 1 x exhaust filter S, with splash guard and change indicator
Set 13
5 L
S50