missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HPS5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13901-1-AP-20UL
This item is not returnable.
View return policy
Description
May regulate the synthesis and function of lysosomes and of highly specialized organelles, such as melanosomes and platelet dense granules. Regulates intracellular vesicular trafficking in fibroblasts. May be involved in the regulation of general functions of integrins.Specifications
HPS5 | |
Polyclonal | |
Unconjugated | |
HPS5 | |
AIBP63, Hermansky Pudlak syndrome 5, HPS5, KIAA1017, Ru2, Ruby eye protein 2 homolog | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11234, 246694, 308598 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot | |
0.43 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P59438, Q9UPZ3 | |
HPS5 | |
HPS5 Fusion Protein Ag4867 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction