Learn More
Thermo Scientific™ hPSC Scorecard Assay Service
hPSC Scorecard Assay Service is a fast and cost-effective alternative to teratoma formation for verification of trilineage differentiation potential in human pluripotent stem cell lines.
Brand:  Thermo Scientific™ A16179
Product Code. 18745395
  • 334.00 € / Each
Explore more special offers
Description
Description
  • A fast and cost-effective alternative to teratoma formation for verification of trilineage differentiation potential in human pluripotent stem cell lines
  • This service leverages the TaqMan hPSC Scorecard Panel (A15872), consisting of 94 predefined TaqMan Gene Expression assays (including endogenous controls) dried-down in the wells
  • The panel consists of genes associated with the following developmental states: pluripotency, ectoderm, endoderm, and mesoderm
The hPSC Scorecard Assay Service offers:
  • Fast data delivery – turnaround time of four weeks or less
  • Convenient sample prep – just send a frozen cell pellet
  • Quantitative results – compare scores to reference set of well-characterized pluripotent stem cell lines
  • Peer endorsement – cited >240 times in scientific literature
  • Exceptional support – a dedicated project manager for frequent project communication
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1 sample
hPSC Scorecard Assay Service
Research Use Only
Human
3 Weeks
Applied Biosystems™, QuantStudio™
Stem Cell Researchers
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ hPSC Scorecard Assay Service > 1 Sample