Learn More
Cytiva HRP labelled Protein A
Protein A conjugated with Horseradish peroxidase for detection of immunoglobulins via affinity to the Fc region - typically IgG from many mammalian species
Brand:  Cytiva NA9120-1ML
Product Code. 10400325
  • 608.00 € / 1mL
Estimated Shipment: 10-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
For use in immunodetection and in situ hybridization applications.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Protein A HRP
For detection of immunoglobulins via affinity to the Fc region - typically IgG from many mammalian species
Complete packsize
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Signal Word
  • Warning
Hazard Category
  • Respiratory sensitiser Category 1