missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IKA™ HS 260 Basic

Compact, flat reciprocal shaker with ideal swivel motion, for a maximum shaking weight of 7.5 kg.
Brand: IKA™ 0003066602
This item is not returnable.
View return policy
Description
- Electronic adjustment of speed and timer
- LED display for speed and time adjustment
- Wide range of attachment combinations makes it possible to use almost all shapes and sizes of vessels

Specifications
0 to 300 rpm | |
5°C to 50°C | |
UK Plug | |
420 x 360 x 100 mm | |
5 to 50 min. |
100/120 V, 220/240 V | |
Reciprocating Shaker | |
50/60 Hz | |
LED line |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
IKA™ HS 260 Basic > Plug Type: UK Plug
Spot an opportunity for improvement?Share a Content Correction