missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trajan™ HT GC Septa
HT GC septa is a bleed and temperature optimized material
68.80 € - 211.00 €
Product Code | Brand | Diameter (Metric) Outer | Quantity | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Outer | Quantity | Price | Quantity & Availability | ||||||
12975721
|
Trajan™
041898 |
11 mm | 25/Pk. |
68.80 €
Pack of 25 |
Estimated Shipment: 30-04-2025
Log in to see stock. |
||||||
12925731
|
Trajan™
041897 |
9.5 mm | 25/Pk. |
68.80 €
Pack of 25 |
Estimated Shipment: 06-05-2025
Log in to see stock. |
||||||
12905731
|
Trajan™
0418990 |
17 mm | 25/Pk. |
86.55 €
Pack of 25 |
Estimated Shipment: 06-05-2025
Log in to see stock. |
||||||
12796377
|
Trajan™
041881 |
9.5 mm | 100/Pk. | N/A | N/A | N/A | |||||
Description
- Bleed and temperature optimized septa
- Reliable seal against carrier gas pressure in the inlet
- No bleed or contamination into the GC system
- Maximum operating temperature of 400°C (330°C for 17mm size)
Specifications
BTO Silicone |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Trajan™ HT GC Septa