missing translation for 'onlineSavingsMsg'
Learn More

HT Supplement (50X)

HT Supplement (50X)

Brand:  Gibco 41065012

Additional Details : Weight : 0.10000kg

Product Code. 11544476

  • 26.65 € / 100mL
In Stock
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Gibco™ HT Supplement is a liquid mixture of sodium hypoxanthine (5 mM) and thymidine (0.8 mM). HT-supplemented medium is suitable for post-selection rescue to overcome the effects of residual intracellular aminopterin. Once the de novo biosynthesis pathway for nucleosides has been reestablished, HT supplementation can be discontinued.

cGMP Manufacturing and Quality System
Gibco™ HT Supplement is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer a comparable Gibco™ HT Supplement product made in our Grand Island facility (11067-030). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
Specifications

Specifications

Storage conditions: 2-8°C
Shipping conditions: Ambient
Shelf life: 18 months from date of manufacture
50 X
HT Supplement
Sustainable packaging
Room Temperature
Mammalian Cells
Liquid
Sterile-filtered
9 to 11
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.