missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HTBA, MedChemExpress
MedChemExpress HTBA(3-Hydroxy-2,4,6-triiodobenzoic acid) for your research needs.
79.65 € - 95.85 €
Specifications
Chemical Name or Material | HTBA |
---|---|
CAS | 53279-72-4 |
Color | Off-White |
Purity Grade Notes | Research |
Molecular Formula | C7H3I3O3 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16451796
|
Medchem Express
HY-15919 |
500 mg |
95.85 €
500mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16436996
|
Medchem Express
HY-15919 |
100 mg |
79.65 €
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HTBA(3-Hydroxy-2,4,6-triiodobenzoic acid) for your research needs.Specifications
HTBA | |
Off-White | |
C7H3I3O3 | |
O=C(O)C1=C(I)C=C(I)C(O)=C1I | |
515.81 | |
Research |
53279-72-4 | |
Research | |
DMSO : ≥ 33 mg/mL (63.98 mM) | |
515.81 | |
99.61% | |
Solid |
Safety and Handling
H302∣H315∣H318∣H335
missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Research purposes only