missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human AID Synthetic Peptide
Synthetic Peptide
Brand: Invitrogen™ PEP0131
This item is not returnable.
View return policy
Description
AID Synthetic Peptide, PEP-0131, from Invitrogen.
This gene encodes a RNA-editing deaminase that is a member of the cytidine deaminase family. The protein is involved in somatic hypermutation, gene conversion, and class-switch recombination of immunoglobulin genes. Defects in this gene are the cause of autosomal recessive hyper-IgM immunodeficiency syndrome type 2 (HIGM2).
Specifications
57379 | |
purified | |
Human AID | |
-20°C | |
Liquid | |
Blocking Assay, Control | |
No | |
Peptide | |
RUO - research use only |
Human | |
Q9GZX7 | |
200 μg/mL | |
Synthetic Peptide | |
PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 | |
AICDA | |
0.02% sodium azide | |
50 μg | |
PBS |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Invitrogen™ Human AID Synthetic Peptide > 50 μg; Unlabeled
Spot an opportunity for improvement?Share a Content Correction