missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human APC11 Synthetic Peptide

Synthetic Peptide

Brand:  Invitrogen™ PEP1069

94.00 EUR valid until 2025-03-21
Use promo code "24111" to get your promotional price.


Product Code. 15265069

  • 141.00 € / 50µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Alert:

To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price

This item is not returnable. View return policy

Description

Description

APC11 Synthetic Peptide, PEP-1069, from Invitrogen.

Together with the cullin protein ANAPC2, constitutes the catalytic component of the anaphase promoting complex/cyclosome (APC/C), a cell cycle-regulated E3 ubiquitin ligase that controls progression through mitosis and the G1 phase of the cell cycle. The APC/C complex acts by mediating ubiquitination and subsequent degradation of target proteins: it mainly mediates the formation of 'Lys-11'-linked polyubiquitin chains and, to a lower extent, the formation of 'Lys-48'- and 'Lys-63'-linked polyubiquitin chains. May recruit the E2 ubiquitin-conjugating enzymes to the complex. [UniProt]
TRUSTED_SUSTAINABILITY
Specifications

Specifications

51529
purified
Human APC11
-20°C
Liquid
Blocking Assay, Control
No
Peptide
RUO - research use only
Human
Q9NYG5
200 μg/mL
Synthetic Peptide
PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
ANAPC11
0.02% sodium azide
50 μg
PBS
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.