missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human APC13 Synthetic Peptide
Synthetic Peptide
Brand: Invitrogen™ PEP1070
This item is not returnable.
View return policy
Description
APC13 Synthetic Peptide, PEP-1070, from Invitrogen.
This peptide corresponds to 17 amino acids near the center of human APC13.PEP-1070 can be used as a blocking peptide with polyclonal antibody PA5-20956.Specifications
25847 | |
purified | |
Human APC13 | |
-20°C | |
Liquid | |
Blocking Assay, Control | |
No | |
Peptide | |
RUO - research use only |
Human | |
Q9BS18 | |
200 μg/mL | |
Synthetic Peptide | |
PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 | |
ANAPC13 | |
0.02% sodium azide | |
50 μg | |
PBS |