Learn More
Gibco™ Human APRIL Recombinant Protein, PeproTech®
Description
310-10C-01M will be provided as 20 x 50 μg (310-10C-50UG). 310-10C-500 will be provided as 10 x 50 μg (310-10C-50UG). 310-10C-250 will be provided as 5 x 50 μg (310-10C-50UG). 310-10C-100 will be provided as 2 x 50 μg (310-10C-50UG) Recombinant human APRIL is a soluble 16.3 kDa protein, consisting of 146 amino acid residues. *Human APRIL (Catalog Number 310-10C) has replaced Human APRIL (Catalog Number 310-10). This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
Specifications
Specifications
| Accession Number | O75888 |
| For Use With (Application) | Control, Bioactivity |
| Formulation | Protein with no preservative |
| Gene ID (Entrez) | 8741 |
| Molecular Weight (g/mol) | 16.3 kDa |
| Name | Human APRIL |
| Purification Method | Purified |
| Quantity | 50 μg |
| Source | Insect cells |
| Storage Requirements | -20°C |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.