missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems Human Biomarker Premixed Magnetic Luminex™ Performance Assay Kit A

Luminex bead-based multiplex assays are designed to provide accurate, reproducible results for every target analyte.

1585.00 € - 2945.00 €

Specifications

Species Human
Binding Target Multiplex Cytokine Assay Kits
For Use With (Application) ELISA
Content And Storage Store the unopened product at 2 - 8 °C. Do not use past expiration date.
View More Specs

Products 5
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30002081
View Documents
R&D Systems
FCSTM13-04
4 Plex
1810.00 €
4 sets
Please to purchase this item. Need a web account? Register with us today!
30002083
View Documents
R&D Systems
FCSTM13-03
3 Plex
1585.00 €
3 sets
Please to purchase this item. Need a web account? Register with us today!
30002076
View Documents
R&D Systems
FCSTM13-09
9 Plex
2945.00 €
9 sets
Please to purchase this item. Need a web account? Register with us today!
30002077
View Documents
R&D Systems
FCSTM13-08
8 Plex
2720.00 €
8 sets
Please to purchase this item. Need a web account? Register with us today!
30002080
View Documents
R&D Systems
FCSTM13-06
6 Plex
2265.00 €
6 sets
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

Human
Multiplex Cytokine Assay Kits
ELISA
Store the unopened product at 2 - 8 °C. Do not use past expiration date.
Videos
SDS
Documents

Documents

Product Certifications