Learn More
Novus Biologicals™ Human Bladder Whole Tissue Lysate (Adult Whole Normal)
Brand:  Novus Biologicals™ NB820-59173
Product Code. 18067463
  • 315.00 € / 1mg
Explore more special offers
Description
Description
The Human Bladder Whole Tissue Lysate (Adult Whole Normal) is derived from human. It has been validated for the following applications: Western Blot, Immunoprecipitation.
Specifications
Specifications
Western Blot
1 mg
Total Protein - Human Adult Normal Tissue: Bladder
Normal
Human
Human Bladder Whole Tissue Lysate (Adult Normal)
Adult
Digestive/Waste
HEPES (pH 7.9), MgCl2, KCl, EDTA, Sucrose, Glycerol, Sodium deoxycholate, NP-40, and a cocktail of protease inhibitors
Whole
Human
5 mg/mL
-80°C. Avoid freeze-thaw cycles.
Native