missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human BMP-4 Recombinant Protein

Recombinant Human Bone Morphogenetic Protein 4 (BMP4) is a bioactive protein intended for use in cell culture applications

Brand:  Gibco™ PHC9531

 View more versions of this product

Product Code. 11310852

  • 2340.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Human BMP-4 Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN

BMP4 is a member of the bone morphogenetic protein family which is part of the transforming growth factor-beta superfamily. The superfamily includes large families of growth and differentiation factors. Bone morphogenetic proteins were originally identified by an ability of demineralized bone extract to induce endochondral osteogenesis in vivo in an extraskeletal site. This particular family member plays an important role in the onset of endochondral bone formation in humans.
  • High purity: no interference from other proteins or contaminants
  • High biological activity: more results with less protein
  • Proven compatibility: Gibco proteins bioassayed with Gibco media
  • High purity recombinant bioactive protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein
  • High biological activity: in-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, anti-viral, and protease assays; specific activity is determined by its ability to induce alkaline phosphatase production by ATDC-5 cells

Shipping Condition: Dry Ice.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Cell Culture
Gel-Purified
BMP-4
Human
Carrier-Free
Induction of alkaline phosphatase in ATDC5 cells
Dry Ice
Growth Factors & Receptors
Lyophilized
Bone Research, Stem Cell Research, Differentiation
95 %
34 kDa
100 μg
116 aa
Store in freezer (-5 to -30°C).
2 ng/ml
< 1 EU/ml
Mammalian
Recombinant, Ligand
BMPs (Bone Morphogenetic Proteins) & Related Factors
Gibco™
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.