missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human BMP-4 Recombinant Protein, PeproTech®

Product Code. p-7246203
Click to view available options
Quantity:
10 μg
100 μg
2 μg
2 x 500 μg
250 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
250µg
2µg
500µg
50µg
Pack of 2
This item is not returnable. View return policy

Product Code. 17811033

Brand: Gibco™ 12005ET100UG

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Recombinant Protein

120-05ET-1MG will be provided as 2 x 500 μg (120-05ET-500UG). PeproTech's E.coli-derived BMP-4 is a fully active homodimeric protein consisting of two 106 amino acid subunits, which correspond to amino acids 303-408 of the full length BMP-4 precursor. The calculated molecular weight of Recombinant Human BMP-4 (E.coli-derived) is 23.9 kDa. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

The protein encoded by this gene is a member of the bone morphogenetic protein family which is part of the transforming growth factor-beta superfamily. The superfamily includes large families of growth and differentiation factors. Bone morphogenetic proteins were originally identified by an ability of demineralized bone extract to induce endochondral osteogenesis in vivo in an extraskeletal site. This particular family member plays an important role in the onset of endochondral bone formation in humans, and a reduction in expression has been associated with a variety of bone diseases, including the heritable disorder Fibrodysplasia Ossificans Progressiva. Alternative splicing in the 5' untranslated region of this gene has been described and three variants are described, all encoding an identical protein.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P12644
For Use With (Application) Control, Western Blot Control, Bioactivity
Formulation Protein with no preservative
Gene ID (Entrez) 652
Molecular Weight (g/mol) 23.9 kDa
Name Human BMP-4
Purification Method Purified
Quantity 100 μg
Source E. coli
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <1 EU/μg
Gene Alias BMP; BMP2B; BMP-2B; Bmp2b1; Bmp2b-1; Bmp4; Bmp-4; BOMPR4A; Bone morphogenetic protein; Bone morphogenetic protein 2B; bone morphogenetic protein 4; bone morphogenetic protein 4 preproprotein; DVR4; Dvr-4; MCOPS6; OFC11; ZYME
Common Name BMP4
Gene Symbol BMP4
Biological Activity Determined by its ability to induce alkaline phosphatase production by ATDC-5 cells. The expected ED50 for this effect is 5-10 ng/ml
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence KKNKNCRRHS LYVDFSDVGW NDWIVAPPGY QAFYCHGDCP FPLADHLNST NHAIVQTLVN SVNSSIPKAC CVPTELSAIS MLYLDEYDKV VLKNYQEMVV EGCGCR
Content And Storage -20°C
Activity Determined by its ability to induce alkaline phosphatase production by ATDC-5 cells. The expected ED50 for this effect is 5-10 ng/ml.
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.