Learn More
Novus Biologicals™ Human Brain Amygdala Whole Tissue Lysate (Adult Whole Alzheimer's)
Brand:  Novus Biologicals™ NB820-59364
Product Code. 18254042
  • 688.00 € / 1mg
Explore more special offers
Description
Description
The Human Brain Amygdala Whole Tissue Lysate (Adult Whole Alzheimer's) is derived from human. It has been validated for the following applications: Western Blot, Immunoprecipitation.
Specifications
Specifications
Western Blot
1 mg
Total Protein - Alzheimers Disease: Brain: Amygdala
Alzheimer's
Human
Human Brain Amygdala Whole Tissue Lysate (Adult Alzheimer's)
Adult
Nervous
HEPES (pH 7.9), MgCl2, KCl, EDTA, Sucrose, Glycerol, Sodium deoxycholate, NP-40, and a cocktail of protease inhibitors
Whole
Human
5 mg/mL
-80°C. Avoid freeze-thaw cycles.
Native