Learn More
Novus Biologicals™ Human Brain Cerebellum Whole Tissue Lysate (Adult Whole Normal)
Applications: Western Blot, Immunoprecipitation
Brand:  Novus Biologicals™ NB820-59180
Product Code. 18771614
  • 351.00 € / 1mg
Explore more special offers
Description
Description
The Human Brain Cerebellum Whole Tissue Lysate (Adult Whole Normal) is derived from human. It has been validated for the following applications: Western Blot, Immunoprecipitation.
Specifications
Specifications
Western Blot
1 mg
Total Protein - Human Adult Normal Tissue: Brain: Cerebellum (left)
Normal
Human
Human Brain Cerebellum Whole Tissue Lysate (Adult Normal)
Adult
Brain
HEPES (pH 7.9), MgCl2, KCl, EDTA, Sucrose, Glycerol, Sodium deoxycholate, NP-40, and a cocktail of protease inhibitors
Whole
Human
5 mg/mL
-80°C. Avoid freeze-thaw cycles.
Native