Learn More
Novus Biologicals™ Human Breast Tissue MicroArray (Tumor)
The panel contains 5 to 8 different tissues on one slide.
Brand:  Novus Biologicals™ NBP2-78087
Product Code. 18609080
  • 544.00 € / Pack of 5
Explore more special offers
Specifications
5 Slides
Human
Please bake slides at 60C for 30 minutes before use.