missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human Btk Synthetic Peptide
Synthetic Peptide
Brand: Invitrogen™ PEP0203
This item is not returnable.
View return policy
Description
BTK Synthetic Peptide, PEP-0203, from Invitrogen.
Bruton tyrosine kinase (BTK) is a cytoplasmic tyrosine kinase belonging to the SRC-related TEC subfamily of tyrosine kinases. Mutations in the BTK gene have been linked to severe developmental blocks in human B-cell ontogeny and immunodeficiency disorders. It has recently been shown to interact with members of the toll-like receptor (TLR) family such as TLR4, 6, 8, and 9. The TLRs are critical molecules in both the innate and adaptive immunity and can recognize diverse microbial pathogens. BTK has also been shown to interact with key proteins involved in TLR4 signal transduction such as MyD88, TIRAP, and IRAK, but not TRAF-6, suggesting that BTK is involved in lipopolysaccharide signal transduction.
Specifications
695 | |
purified | |
Human Btk | |
-20°C | |
Liquid | |
Blocking Assay, Control | |
No | |
Peptide | |
RUO - research use only |
Human | |
Q06187 | |
200 μg/mL | |
Synthetic Peptide | |
PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 | |
BTK | |
0.02% sodium azide | |
50 μg | |
PBS |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Invitrogen™ Human Btk Synthetic Peptide > 50 μg; Unlabeled
Spot an opportunity for improvement?Share a Content Correction