Learn More
Invitrogen™ Human Carbonic Anhydrase III Recombinant Protein
Recombinant Protein
Brand:  Invitrogen™ RP75524
Product Code. 11584211
  • 275.00 € / 10µg
Explore more special offers
Description
Description
CAIII (human) Recombinant Protein, RP-75524, from Invitrogen.
This product contains purified recombinant human CAIII protein with an N-terminal hexahistidine tag. This product may be used as a positive control in many immunological assays.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
P07451
PBS with 50% glycerol and no preservative
Human Carbonic Anhydrase III
E. coli
-20°C, Avoid Freeze/Thaw Cycles
Carbonic Anhydrase III
CA3
Unconjugated
E. coli
Liquid
purified
Control, Western Blot
761
10 μg
N/A
RUO
Human Carbonic Anhydrase III
Human
Hu
Recombinant
Recombinant
Product Suggestions