Learn More
Invitrogen™ Human Carbonic Anhydrase VB (aa 176-279) Control Fragment Recombinant Protein
Recombinant Protein
Brand:  Invitrogen™ RP92233
Product Code. 30206187
  • 259.00 € / 100µL
Explore more special offers
Description
Description
Carbonic anhydrases (CAs) are members of a large family of zinc metalloenzymes responsible for catalyzing the reversible hydration of carbon dioxide.CAs show extensive diversity in their distribution and subcellular localization.They are involved in a variety of biological processes, including calcification, bone resorption, respiration, acid-base balance and the formation of aqueous humor, saliva, gastric juice and cerebrospinal fluid. CA VB, also known as carbonate dehydratase VB, is one of two isoforms of CA V. It localizes to the mitochondria and is involved in metabolic processes. CA VB is predominantly expressed in heart, pancreas, lung, placenta, kidney and skeletal muscle. It exhibits highest homology with family member CA VA (the second isoform of CA V); however, unlike CA VA, it is not expressed in the liver, suggesting that it plays a significantly different physiological role.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Q9Y2D0
Blocking Assay, Control
11238
100 μL
RUO
CA5B
Human
GLAVIGVFLKLGKHHKELQKLVDTLPSIKHKDALVEFGSFDPSCLMPTCPDYWTYSGSLTTPPLSESVTWIIKKQPVEVDHDQLEQFRTLLFTSEGEKEKRMVD
Liquid
≥5.0 mg/mL
1M urea, PBS with no preservative; pH 7.4
Carbonic Anhydrase VB
-20° C, Avoid Freeze/Thaw Cycles
7330410H16Rik; Ca5b; Car5b; Carbonate dehydratase VB; carbonic anhydrase 5B; carbonic anhydrase 5b, mitochondrial; Carbonic anhydrase VB; carbonic anhydrase VB, mitochondrial; carbonic dehydratase; CarVb; CAVB; CA-VB; D730005F19Rik
Unconjugated
Recombinant
E. coli