Learn More
Invitrogen™ Human Carbonic Anhydrase VI (aa 119-215) Control Fragment Recombinant Protein
Recombinant Protein
Brand:  Invitrogen™ RP94191
Product Code. 30194647
  • 259.00 € / 100µL
Explore more special offers
Description
Description
The protein encoded by this gene is one of several isozymes of carbonic anhydrase. This protein is found only in salivary glands and saliva and protein may play a role in the reversible hydratation of carbon dioxide though its function in saliva is unknown.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
P23280
Blocking Assay, Control
765
100 μL
RUO
CA6
Human
SEISGSEHTVDGIRHVIEIHIVHYNSKYKSYDIAQDAPDGLAVLAAFVEVKNYPENTYYSNFISHLANIKYPGQRTTLTGLDVQDMLPRNLQHYYTY
Liquid
≥5.0 mg/mL
1M urea, PBS with no preservative; pH 7.4
Carbonic Anhydrase VI
-20° C, Avoid Freeze/Thaw Cycles
CA6; CA-6; Car6; Carbonate dehydratase VI; carbonic anhydrase 6; Carbonic anhydrase VI; carbonic anhydrase VI nirs variant 2; Carbonic anhydrase6; CA-VI; DOC1; GUSTIN; salivary carbonic anhydrase; Secreted carbonic anhydrase
Unconjugated
Recombinant
E. coli